missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-méthoxybenzaldéhyde 98,0+%, TCI America™

Numéro de catalogue. p-7130677
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H06995G 5 g
H069925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H06995G Fournisseur TCI America Code fournisseur H06995G

Identifiants chimiques

CAS 673-22-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00003327
InChI Key WZUODJNEIXSNEU-UHFFFAOYSA-N
Synonym 4-methoxysalicylaldehyde, benzaldehyde, 2-hydroxy-4-methoxy, 4-methoxysalicyaldehyde, 2-hydroxy-4-methoxy-benzaldehyde, o-hydroxy-p-methoxybenzaldehyde, p-anisaldehyde, 2-hydroxy, salicylaldehyde, 4-methoxy, 2-hydroxy-p-anisaldehyde, 2-formyl-5-methoxyphenol, unii-2n395p88lw
PubChem CID 69600
IUPAC Name 2-hydroxy-4-méthoxybenzaldéhyde
SMILES COC1=CC=C(C=O)C(O)=C1

Spécifications

Melting Point 42°C
Quantity 5 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Poudre cristalline
Chemical Name or Material 2-Hydroxy-4-methoxybenzaldehyde
RTECSNumber BZ2810000
TSCA Yes