accessibility menu, dialog, popup

UserName

CAS RN 673-22-3

IUPAC Name: 2-hydroxy-4-methoxybenzaldehyde
Synonyms: 2-hydroxy-4-methoxybenzaldehyde
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: WZUODJNEIXSNEU-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C(O)=C1
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)