missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxyaniline Hydrochloride 98.0+%, TCI America™

Catalog No. A12595G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12595G 5 g
A12591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12595G Supplier TCI America Supplier No. A12595G
Only null left

Chemical Identifiers

CAS 39547-15-4
Molecular Formula C7H10ClNO2
Molecular Weight (g/mol) 175.61
MDL Number MFCD00052002
InChI Key ZXJIQOBYRBTOLH-UHFFFAOYSA-N
Synonym 2-Amino-5-methoxyphenol Hydrochloride, 2-Hydroxy-p-anisidine Hydrochloride
PubChem CID 12210184
IUPAC Name 2-amino-5-methoxyphenol hydrochloride
SMILES Cl.COC1=CC(O)=C(N)C=C1

Specifications

Melting Point 213°C
Quantity 5 g
Formula Weight 175.61
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-4-methoxyaniline Hydrochloride