missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Hydroxy-4',6'-dimethoxy-2-phenylacetophenone 94.0+%, TCI America™
Specifications
Specifications
| Melting Point | 117°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 272.30 |
| Percent Purity | ≥94.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2′-Hydroxy-4′,6′-dimethoxy-2-phenylacetophenone |