missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Hydroxy-4',6'-dimethoxy-2-(4-methoxyphenyl)acetophenone 98.0+%, TCI America™
$162.17 - $486.52
Chemical Identifiers
| CAS | 39604-68-7 |
|---|---|
| Molecular Formula | C17H18O5 |
| Molecular Weight (g/mol) | 302.33 |
| MDL Number | MFCD03934798 |
| InChI Key | ROJOJPXRGHLGTC-UHFFFAOYSA-N |
| Synonym | 2-Hydroxy-4,6-dimethoxyphenyl 4-Methoxybenzyl Ketone |
| PubChem CID | 689039 |
| IUPAC Name | 1-(2-hydroxy-4,6-dimethoxyphenyl)-2-(4-methoxyphenyl)ethan-1-one |
| SMILES | COC1=CC=C(CC(=O)C2=C(O)C=C(OC)C=C2OC)C=C1 |
Chemical Identifiers
| 39604-68-7 | |
| 302.33 | |
| ROJOJPXRGHLGTC-UHFFFAOYSA-N | |
| 689039 | |
| COC1=CC=C(CC(=O)C2=C(O)C=C(OC)C=C2OC)C=C1 |
| C17H18O5 | |
| MFCD03934798 | |
| 2-Hydroxy-4,6-dimethoxyphenyl 4-Methoxybenzyl Ketone | |
| 1-(2-hydroxy-4,6-dimethoxyphenyl)-2-(4-methoxyphenyl)ethan-1-one |
Specifications
| 39604-68-7 | |
| White-Yellow | |
| MFCD03934798 | |
| 2-Hydroxy-4,6-dimethoxyphenyl 4-Methoxybenzyl Ketone | |
| COC1=CC=C(CC(=O)C2=C(O)C=C(OC)C=C2OC)C=C1 | |
| 302.33 | |
| 302.33 | |
| Crystalline Powder |
| 87°C | |
| C17H18O5 | |
| 1 g | |
| ROJOJPXRGHLGTC-UHFFFAOYSA-N | |
| 1-(2-hydroxy-4,6-dimethoxyphenyl)-2-(4-methoxyphenyl)ethan-1-one | |
| 689039 | |
| ≥98.0% (GC) | |
| 2′-Hydroxy-4′,6′-dimethoxy-2-(4-methoxyphenyl)acetophenone |
Safety and Handling
TSCA : No