accessibility menu, dialog, popup

UserName

CAS RN 39604-68-7

IUPAC Name: 1-(2-hydroxy-4,6-dimethoxyphenyl)-2-(4-methoxyphenyl)ethan-1-one
Synonyms: 1-(2-hydroxy-4,6-dimethoxyphenyl)-2-(4-methoxyphenyl)ethanone
Molecular Weight (g/mol): 302.33
Molecular Formula: C17H18O5
InChi Key: ROJOJPXRGHLGTC-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC(=O)C2=C(O)C=C(OC)C=C2OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results