missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-3-phenoxypropyl Acrylate (stabilized with MEHQ) 85.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 222.24 |
| Percent Purity | ≥85.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Hydroxy-3-phenoxypropyl Acrylate (stabilized with MEHQ) |
Safety and Handling
| EINECSNumber | (3)-0565 |
| TSCA | Yes |