missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-nitropyridine 98.0+%, TCI America™

Catalog No. p-7130853
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09995G 5 g
H099925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09995G Supplier TCI America Supplier No. H09995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6332-56-5 | C5H4N2O3 | 140.10 g/mol

Chemical Identifiers

CAS 6332-56-5
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.10
MDL Number MFCD00006270
InChI Key BOAFCICMVMFLIT-UHFFFAOYSA-N
Synonym 2-hydroxy-3-nitropyridine, 3-nitropyridin-2-ol, 3-nitro-2-pyridinol, 3-nitropyridin-2 1h-one, 2 1h-pyridinone, 3-nitro, 3-nitro-2-hydroxypyridine, pyridine, 2-hydroxy-3-nitro, 3-nitro-1,2-dihydropyridin-2-one, 3-nitro-2-oxopyridine, 2-pyridinol, 3-nitro
PubChem CID 22793
IUPAC Name 3-nitro-1,2-dihydropyridin-2-one
SMILES [O-][N+](=O)C1=CC=CNC1=O

Specifications

Melting Point 226°C
Color Yellow
Quantity 5 g
Formula Weight 140.10
Percent Purity ≥98.0% (HPLC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-3-nitropyridine
RTECSNumber UU7718000
TSCA No