accessibility menu, dialog, popup

UserName

ONOHNO

CAS RN 6332-56-5

IUPAC Name: 3-nitro-1,2-dihydropyridin-2-one
Synonyms: 3-nitro-1H-pyridin-2-one
Molecular Weight (g/mol): 140.1
Molecular Formula: C5H4N2O3
InChi Key: BOAFCICMVMFLIT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CNC1=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results