missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methylpyrazine 98.0+%, TCI America™

Catalog No. H15195G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15195G 5 g
H15191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15195G Supplier TCI America Supplier No. H15195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19838-07-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD01646120
InChI Key LDQRWMQHTORUIY-UHFFFAOYSA-N
PubChem CID 88276
IUPAC Name 3-methyl-1H-pyrazin-2-one
SMILES CC1=NC=CNC1=O

Specifications

Melting Point 150°C
Color Yellow
Quantity 5 g
Synonym 2-hydroxy-3-methylpyrazine, 3-methylpyrazin-2-ol, 3-methylpyrazin-2 1h-one, 3-methyl-2-pyrazinol, 3-methyl-2 1h-pyrazinone, unii-s951qmj75e, 2 1h-pyrazinone, 3-methyl, 3-methyl-2-hydroxypyrazine, 2 1h-pyrazinone,3-methyl
Formula Weight 110.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-3-methylpyrazine