accessibility menu, dialog, popup

UserName

CH3NHNO

CAS RN 19838-07-4

IUPAC Name: 3-methyl-1,2-dihydropyrazin-2-one
Synonyms: 3-methyl-1H-pyrazin-2-one
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: LDQRWMQHTORUIY-UHFFFAOYSA-N
SMILES: CC1=NC=CNC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results