missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methoxybenzoic acid, 98+%

Catalog No. p-7043625
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0165806 5 g
AAL0165814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0165806 Supplier Thermo Scientific Chemicals Supplier No. L0165806
Only null left

CAS: 877-22-5 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 877-22-5
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002445
InChI Key AUZQQIPZESHNMG-UHFFFAOYSA-N
Synonym 3-methoxysalicylic acid, o-vanillic acid, acide orthovanillique, benzoic acid, 2-hydroxy-3-methoxy, m-anisic acid, 2-hydroxy, 3-hydroxy-m-anisic acid, unii-3j31y0e2im, 2-hydroxy-3-methoxy-benzoic acid, 3-methoxysalicyclic acid, 2-hydroxy-3-methoxy benzoic acid
PubChem CID 70140
ChEBI CHEBI:68496
IUPAC Name 2-hydroxy-3-methoxybenzoic acid
SMILES COC1=CC=CC(C(O)=O)=C1O

Specifications

Melting Point 151°C to 152°C
Quantity 5 g
Beilstein 2209642
Formula Weight 168.15
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxy-3-methoxybenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 212-888-9
RTECSNumber BZ4840000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only