Learn More
2-Hydroxy-3-methoxybenzaldehyde, 99%
CAS: 148-53-8 | C8H8O3 | 152.15 g/mol
Supplier: Thermo Scientific Chemicals A1567222
Description
o-Vanillin has been used to study the solvent-free reaction between o-vanillin and p-toluidine using NMR, DSC and XRD analysis. It was used in the synthesis of new ligand for Fe(III) and Al(lII). It is also used in the study of mutagenesis and as a synthetic precursor for pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationso-Vanillin has been used to study the solvent-free reaction between o-vanillin and p-toluidine using NMR, DSC and XRD analysis. It was used in the synthesis of new ligand for Fe(III) and Al(lII). It is also used in the study of mutagenesis and as a synthetic precursor for pharmaceuticals.
Solubility
Solubility in methanol is almost transparent. Soluble in alcohol, methanol and dioxane (1g in 10 mL). Insoluble in water.
Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 2-Hydroxy-3-methoxybenzaldehyde | |
| 40°C to 42°C | |
| C8H8O3 | |
| 100 g | |
| Air Sensitive | |
| JJVNINGBHGBWJH-UHFFFAOYSA-N | |
| 2-hydroxy-3-methoxybenzaldehyde | |
| 8991 | |
| 152.15 |
| 148-53-8 | |
| 265°C to 266°C | |
| MFCD00003322 | |
| 471913 | |
| 3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehyde | |
| COC1=CC=CC(C=O)=C1O | |
| 152.15 | |
| CHEBI:78339 | |
| 99% |
Chemical Identifiers
| 148-53-8 | |
| 152.15 | |
| JJVNINGBHGBWJH-UHFFFAOYSA-N | |
| 8991 | |
| 2-hydroxy-3-methoxybenzaldehyde |
| C8H8O3 | |
| MFCD00003322 | |
| 3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehyde | |
| CHEBI:78339 | |
| COC1=CC=CC(C=O)=C1O |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 205-715-3
RTECSNumber : CU6530000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only