missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methoxybenzaldehyde, 99%

Catalog No. AAA1567214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1567214 25 g
AAA1567222 100 g
AAA1567236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1567214 Supplier Thermo Scientific Chemicals Supplier No. A1567214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 148-53-8 | C8H8O3 | 152.15 g/mol

o-Vanillin has been used to study the solvent-free reaction between o-vanillin and p-toluidine using NMR, DSC and XRD analysis. It was used in the synthesis of new ligand for Fe(III) and Al(lII). It is also used in the study of mutagenesis and as a synthetic precursor for pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Vanillin has been used to study the solvent-free reaction between o-vanillin and p-toluidine using NMR, DSC and XRD analysis. It was used in the synthesis of new ligand for Fe(III) and Al(lII). It is also used in the study of mutagenesis and as a synthetic precursor for pharmaceuticals.

Solubility
Solubility in methanol is almost transparent. Soluble in alcohol, methanol and dioxane (1g in 10 mL). Insoluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 148-53-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00003322
InChI Key JJVNINGBHGBWJH-UHFFFAOYSA-N
Synonym 3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehyde
PubChem CID 8991
ChEBI CHEBI:78339
IUPAC Name 2-hydroxy-3-methoxybenzaldehyde
SMILES COC1=CC=CC(C=O)=C1O

Specifications

Melting Point 40°C to 42°C
Boiling Point 265°C to 266°C
Quantity 25 g
Beilstein 471913
Sensitivity Air Sensitive
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material 2-Hydroxy-3-methoxybenzaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 205-715-3
RTECSNumber CU6530000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only