missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-methylbutyric Acid, 95+%, Spectrum™ Chemical

$221.86 - $171,072.29

Chemical Identifiers

CAS 3739-30-8
Molecular Weight (g/mol) 118.13 g/mol
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18608219
View Documents Promotion Details
N/A 5 g Poly Bottle
Each for $221.86
Only null left
 
18608220
View Documents Promotion Details
N/A 25 g Poly Bottle
Each for $635.07
Only null left
 
18608221
View Documents Promotion Details
N/A 100 g Poly Bottle
Each for $1,815.82
Only null left
 
18608222
View Documents Promotion Details
N/A 25 kg Poly Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

3739-30-8
118.13 g/mol

Specifications

3739-30-8
White
72°C to 75°C
95%
Crystalline Powder or Lumps
1
237.6°C
118.13 g/mol
Reagent
2-Hydroxy-2-methylbutyric Acid
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.