missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-methylbutyric Acid, 95+%, Spectrum™ Chemical

Catalog No. 18608219
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
25 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608219 5 g Poly Bottle
18608220 25 g Poly Bottle
18608221 100 g Poly Bottle
18608222 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608219 Supplier Spectrum Chemical Mfg Cor Supplier No. H40365GM
Only null left

Chemical Identifiers

CAS 3739-30-8
Molecular Weight (g/mol) 118.13 g/mol

Specifications

CAS Max % 1
Color White
Boiling Point 237.6°C
Melting Range 72°C to 75°C
Packaging Poly Bottle
Quantity 5 g
Percent Purity 95%
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material 2-Hydroxy-2-methylbutyric Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.