missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-1-naphthoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 167°C |
| Quantity | 25 g |
| Formula Weight | 188.18 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Hydroxy-1-naphthoic Acid |
Safety and Handling
| EINECSNumber | (4)-0398 |
| TSCA | Yes |