missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-1-naphthaldehyde, tech.

CAS: 708-06-5 | C11H8O2 | 172.18 g/mol

$438.90 - $438.90

Chemical Identifiers

CAS 708-06-5
MDL Number MFCD00004005
InChI Key NTCCNERMXRIPTR-UHFFFAOYSA-N
Synonym 2-hydroxy-1-naphthaldehyde, 1-formyl-2-naphthol, 2-hydroxy-1-naphthylaldehyde, 2-hydroxy-1-napthaldehyde, 2-hydroxynaphthaldehyde, 2-naphthol 1-carboxaldehyde, 2-hydroxy-1-naphthalaldehyde, 1-naphthalenecarboxaldehyde, 2-hydroxy, 2-hydroxy-1-naphthalenecarboxaldehyde, 1-hydroxy-2-naphthalenecarboxaldehyde
PubChem CID 12819
IUPAC Name 2-hydroxynaphthalene-1-carbaldehyde
SMILES C1=CC=C2C(=C1)C=CC(=C2C=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC121605000
SDS View Documents
Thermo Scientific Chemicals
121605000
500 g
Each for $438.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

708-06-5
NTCCNERMXRIPTR-UHFFFAOYSA-N
12819
C1=CC=C2C(=C1)C=CC(=C2C=O)O
MFCD00004005
2-hydroxy-1-naphthaldehyde, 1-formyl-2-naphthol, 2-hydroxy-1-naphthylaldehyde, 2-hydroxy-1-napthaldehyde, 2-hydroxynaphthaldehyde, 2-naphthol 1-carboxaldehyde, 2-hydroxy-1-naphthalaldehyde, 1-naphthalenecarboxaldehyde, 2-hydroxy, 2-hydroxy-1-naphthalenecarboxaldehyde, 1-hydroxy-2-naphthalenecarboxaldehyde
2-hydroxynaphthalene-1-carbaldehyde

Specifications

708-06-5
Yellow
Authentic
C11H8O2
MFCD00004005
08,143
Solubility in water: 40mg/l
C1=CC=C2C(=C1)C=CC(=C2C=O)O
172.18
172.18
Technical
2-Hydroxy-1-naphthaldehyde, tech.
>77.0°C
192.0°C (27.0 mmHg)
Plastic bottle
HOC10H6CHO
500 g
2-hydroxy-1-naphthaldehyde, 1-formyl-2-naphthol, 2-hydroxy-1-naphthylaldehyde, 2-hydroxy-1-napthaldehyde, 2-hydroxynaphthaldehyde, 2-naphthol 1-carboxaldehyde, 2-hydroxy-1-naphthalaldehyde, 1-naphthalenecarboxaldehyde, 2-hydroxy, 2-hydroxy-1-naphthalenecarboxaldehyde, 1-hydroxy-2-naphthalenecarboxaldehyde
NTCCNERMXRIPTR-UHFFFAOYSA-N
2-hydroxynaphthalene-1-carbaldehyde
12819
≥93% (on dry substance)
Moist Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-902-0

RTECSNumber : QJ0300000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only