missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-1,4-naphthoquinone 98.0+%, TCI America™

Catalog No. p-7130463
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H028525G 25 g
H0285100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H028525G Supplier TCI America Supplier No. H028525G

Chemical Identifiers

CAS 83-72-7
Molecular Formula C10H6O3
Molecular Weight (g/mol) 174.155
MDL Number MFCD00001678
InChI Key WVCHIGAIXREVNS-UHFFFAOYSA-N
Synonym lawsone, 2-hydroxy-1,4-naphthoquinone, 2-hydroxynaphthalene-1,4-dione, 2-hydroxynaphthoquinone, henna, mehendi, hana, lawson, mendi, flower of paradise
PubChem CID 6755
IUPAC Name 4-hydroxynaphthalene-1,2-dione
SMILES C1=CC=C2C(=C1)C(=CC(=O)C2=O)O

Specifications

Quantity 25 g
Formula Weight 174.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-1,4-naphthoquinone
EINECSNumber (4)-0375
RTECSNumber QL8200000
TSCA Yes