missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-1,4-naphthoquinone, 98+%

CAS: 83-72-7 | C10H6O3 | 174.155 g/mol

Supplier:  Thermo Scientific Chemicals A1188018

 View more versions of this product

Catalog No. AAA1188018


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Hydroxy-1,4-naphthoquinone is used for preparing decorative hair and skin dyes. 2-Hydroxy-1,4-naphthoquinone also demonstrates antimicrobial and antioxidant effects. It also suppress the formation of hydrogen peroxide and superoxide radical anion by aldehyde oxidase-catalyzed reactions.

Solubility
Soluble in water. 2 g/L at 20°C

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
Specifications

Specifications

2-Hydroxy-1,4-naphthoquinone
∼193°C (decomposition)
MFCD00001678
1565260
lawsone, 2-hydroxy-1,4-naphthoquinone, 2-hydroxynaphthalene-1,4-dione, 2-hydroxynaphthoquinone, henna, mehendi, hana, lawson, mendi, flower of paradise
WVCHIGAIXREVNS-UHFFFAOYSA-N
4-hydroxynaphthalene-1,2-dione
6755
≥98%
83-72-7
C10H6O3
50 g
14,5393
Soluble in water. 2g/L at 20°C
C1=CC=C2C(=C1)C(=CC(=O)C2=O)O
174.155
174.16

Chemical Identifiers

83-72-7
174.155
WVCHIGAIXREVNS-UHFFFAOYSA-N
6755
C1=CC=C2C(=C1)C(=CC(=O)C2=O)O
C10H6O3
MFCD00001678
lawsone, 2-hydroxy-1,4-naphthoquinone, 2-hydroxynaphthalene-1,4-dione, 2-hydroxynaphthoquinone, henna, mehendi, hana, lawson, mendi, flower of paradise
4-hydroxynaphthalene-1,2-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H302-H319
Suspected of causing genetic defects.
Harmful if swallowed.
Causes serious eye irritation.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H341

EINECSNumber : 201-496-3

RTECSNumber : QL8200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only