missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazinopyrazine, 98%

Catalog No. p-7055491
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6408603 1 g
AAH6408606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6408603 Supplier Thermo Scientific Chemicals Supplier No. H6408603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54608-52-5 | C4H6N4 | 110.12 g/mol

Triazolo[4,3-a]pyrazine 7-oxide, 3-β-D-ribofuranosyl-1,2,4-triazolo[4,3-a]pyrazine, monohydrazones of pyridine, pyrazine, phenylglyoxal were synthesised from 2-Hydrazinopyrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triazolo[4,3-a]pyrazine 7-oxide, 3-β-D-ribofuranosyl-1,2,4-triazolo[4,3-a]pyrazine, monohydrazones of pyridine, pyrazine, phenylglyoxal were synthesised from 2-Hydrazinopyrazine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 54608-52-5
Molecular Formula C4H6N4
Molecular Weight (g/mol) 110.12
MDL Number MFCD04114555
InChI Key IVRLZJDPKUSDCF-UHFFFAOYSA-N
Synonym 2-hydrazinylpyrazine, 2-hydrazinopyrazine, pyrazin-2-yl-hydrazine, 1-pyrazin-2-yl hydrazine, hydrazinopyrazine, 2-pyrazin-2-yl hydrazine, 2 1h-pyrazinone, hydrazone, hydrazino pyrazine, pyrazinyl hydrazine, 3-hydrazinopyrazine
PubChem CID 1487823
IUPAC Name pyrazin-2-ylhydrazine
SMILES C1=CN=C(C=N1)NN

Specifications

Melting Point 109°C to 113°C
Color Yellow
Quantity 1 g
Beilstein 606630
Solubility Information Slightly soluble in water.
Formula Weight 110.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Hydrazinopyrazine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only