CAS RN 54608-52-5
IUPAC Name:
2-hydrazinylpyrazine
Synonyms:
2-hydrazinylpyrazine
Molecular Weight (g/mol):
110.12
Molecular Formula:
C4H6N4
InChi Key:
IVRLZJDPKUSDCF-UHFFFAOYSA-N
SMILES:
NNC1=NC=CN=C1
1
–
1
of
1
results
1
Promotions Available
2-Hydrazinopyrazine, 98%
2-Hydrazinopyrazine, 98%
CAS: 54608-52-5 Molecular Formula: C4H6N4 Molecular Weight (g/mol): 110.12 MDL Number: MFCD04114555 InChI Key: IVRLZJDPKUSDCF-UHFFFAOYSA-N Synonym: 2-hydrazinylpyrazine,2-hydrazinopyrazine,pyrazin-2-yl-hydrazine,1-pyrazin-2-yl hydrazine,hydrazinopyrazine,2-pyrazin-2-yl hydrazine,2 1h-pyrazinone, hydrazone,hydrazino pyrazine,pyrazinyl hydrazine,3-hydrazinopyrazine PubChem CID: 1487823 IUPAC Name: pyrazin-2-ylhydrazine SMILES: C1=CN=C(C=N1)NN