missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazino-2-imidazoline Hydrobromide 98.0+%, TCI America™

Catalog No. p-7131090
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15231G 1 g
H15235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15231G Supplier TCI America Supplier No. H15231G
Only null left

Chemical Identifiers

CAS 55959-84-7
Molecular Formula C3H9BrN4
Molecular Weight (g/mol) 181.037
MDL Number MFCD00012694
InChI Key HIJRJVVSABWXKG-UHFFFAOYSA-N
Synonym 2-hydrazino-2-imidazoline hydrobromide, 2-hydrazinyl-4,5-dihydro-1h-imidazole hydrobromide, 2-hydrazino-delta-2-imidazoline hydrobromide, 4,5-dihydroimidazol-2-ylhydrazine hydrobromide, 4,5-dihydro-2-hydrazino-1h-imidazole hydrobromide, 2-hydrazino-4,5-dihydro-1h-imidazole hydrobromide, 4,5-dihydro-1h-imidazol-2-ylhydrazine hydrobromide, 4,5-dihydro-1h-imidazol-2-yl-hydrazine hydrobromide, 4,5-dihydro-1h-imidazol-2-yl-hydrazine, acmc-1av1k
PubChem CID 3085099
IUPAC Name 4,5-dihydro-1H-imidazol-2-ylhydrazine;hydrobromide
SMILES C1CN=C(N1)NN.Br

Specifications

Melting Point 185°C
Color White
Quantity 1 g
Formula Weight 181.04
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydrazino-2-imidazoline Hydrobromide