accessibility menu, dialog, popup

UserName

CAS RN 55959-84-7

IUPAC Name: hydrogen 2-hydrazinyl-4,5-dihydro-1H-imidazole bromide
Synonyms: hydrogen 2-hydrazinyl-4,5-dihydro-1H-imidazole bromide
Molecular Weight (g/mol): 181.04
Molecular Formula: C3H9BrN4
InChi Key: HIJRJVVSABWXKG-UHFFFAOYSA-N
SMILES: [H+].[Br-].NNC1=NCCN1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)