missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazino-2-imidazoline hydrobromide, 98%

Catalog No. p-7025066
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2038688 2.5 g
AAB2038609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2038688 Supplier Thermo Scientific Chemicals Supplier No. B2038688
Only null left

CAS: 55959-84-7 | C3H9BrN4 | 181.037 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55959-84-7
Molecular Formula C3H9BrN4
Molecular Weight (g/mol) 181.037
MDL Number MFCD00012694
InChI Key HIJRJVVSABWXKG-UHFFFAOYSA-N
Synonym 2-hydrazino-2-imidazoline hydrobromide, 2-hydrazinyl-4,5-dihydro-1h-imidazole hydrobromide, 2-hydrazino-delta-2-imidazoline hydrobromide, 4,5-dihydroimidazol-2-ylhydrazine hydrobromide, 4,5-dihydro-2-hydrazino-1h-imidazole hydrobromide, 2-hydrazino-4,5-dihydro-1h-imidazole hydrobromide, 4,5-dihydro-1h-imidazol-2-ylhydrazine hydrobromide, 4,5-dihydro-1h-imidazol-2-yl-hydrazine hydrobromide, 4,5-dihydro-1h-imidazol-2-yl-hydrazine, acmc-1av1k
PubChem CID 3085099
IUPAC Name 4,5-dihydro-1H-imidazol-2-ylhydrazine;hydrobromide
SMILES C1CN=C(N1)NN.Br

Specifications

Melting Point 181°C to 185°C
Quantity 2.5 g
Sensitivity Hygroscopic
Formula Weight 181.04
Percent Purity 98%
Chemical Name or Material 2-Hydrazino-2-imidazoline hydrobromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 259-921-3
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only