missing translation for 'onlineSavingsMsg'
Learn More

2-Hexynoic acid, 96%

Catalog No. p-7043753
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0252603 1 g
AAL0252606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0252603 Supplier Thermo Scientific Chemicals Supplier No. L0252603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 764-33-0 | C6H8O2 | 112.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 764-33-0
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00014371
InChI Key AKYAUBWOTZJUBI-UHFFFAOYSA-N
Synonym 2-hexynoic acid, hexynoic acid, acmc-1bezk, akyaubwotzjubi-uhfffaoysa
PubChem CID 324380
IUPAC Name hex-2-ynoic acid
SMILES CCCC#CC(=O)O

Specifications

Melting Point 23°C to 26°C
Boiling Point 110°C to 112°C (10 mmHg)
Refractive Index 1.459
Quantity 1 g
UN Number UN3261
Beilstein 1744983
Formula Weight 112.13
Percent Purity 96%
Chemical Name or Material 2-Hexynoic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only