accessibility menu, dialog, popup

UserName

CH3HOO

CAS RN 764-33-0

IUPAC Name: hex-2-ynoic acid
Synonyms: hex-2-ynoic acid
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: AKYAUBWOTZJUBI-UHFFFAOYSA-N
SMILES: CCCC#CC(O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)