missing translation for 'onlineSavingsMsg'
Learn More

2-Hexyne, 98+%

Catalog No. AAB2240518
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2240518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2240518 Supplier Thermo Scientific Chemicals Supplier No. B2240518

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 764-35-2 | C6H10 | 82.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 764-35-2
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00009470
InChI Key MELUCTCJOARQQG-UHFFFAOYSA-N
Synonym 2-hexyne, methyl propyl acetylene, methylpropylacetylene, acmc-209p3f, 1-methyl-2-propylacetylene, c3h7c.$.cch3
PubChem CID 33629
IUPAC Name hex-2-yne
SMILES CCCC#CC

Specifications

Melting Point -90°C
Density 0.736
Boiling Point 84°C to 86°C
Flash Point −10°C (14°F)
Odor Characteristic
Refractive Index 1.414
Quantity 50 g
UN Number UN3295
Beilstein 1731542
Formula Weight 82.15
Percent Purity ≥98%
Chemical Name or Material 2-Hexyne
Show More Show Less
Hazard Category H225-H304-H315-H319-H335
Hazard Statement GHS H Statement
H225-H304-H315-H319-H335
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 212-117-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only