CAS RN 764-35-2
IUPAC Name:
hex-2-yne
Synonyms:
2-hexyne
Molecular Weight (g/mol):
82.15
Molecular Formula:
C6H10
InChi Key:
MELUCTCJOARQQG-UHFFFAOYSA-N
SMILES:
CCCC#CC
1
–
2
of
2
results
1
2-Hexyne, 98+%
CAS: 764-35-2 Molecular Formula: C6H10 Molecular Weight (g/mol): 82.146 MDL Number: MFCD00009470 InChI Key: MELUCTCJOARQQG-UHFFFAOYSA-N Synonym: 2-hexyne,methyl propyl acetylene,methylpropylacetylene,acmc-209p3f,1-methyl-2-propylacetylene,c3h7c.$.cch3 PubChem CID: 33629 IUPAC Name: hex-2-yne SMILES: CCCC#CC
2
2-Hexyne 98.0+%, TCI America™
CAS: 764-35-2 Molecular Formula: C6H10 Molecular Weight (g/mol): 82.146 MDL Number: MFCD00009470 InChI Key: MELUCTCJOARQQG-UHFFFAOYSA-N Synonym: 2-hexyne,methyl propyl acetylene,methylpropylacetylene,acmc-209p3f,1-methyl-2-propylacetylene,c3h7c.$.cch3 PubChem CID: 33629 IUPAC Name: hex-2-yne SMILES: CCCC#CC