missing translation for 'onlineSavingsMsg'
Learn More

2-Hexyldecan-1-amine 98.0+%, TCI America™

Catalog No. p-7131136
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16341ML 1 mL
H16345ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H16341ML Supplier TCI America Supplier No. H16341ML
Only null left

Chemical Identifiers

CAS 62281-05-4
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.463
InChI Key HEGUXAZWCQVLPV-UHFFFAOYSA-N
PubChem CID 15335006
IUPAC Name 2-hexyldecan-1-amine
SMILES CCCCCCCCC(CCCCCC)CN

Specifications

Color Yellow
Boiling Point 150°C
Quantity 1 mL
UN Number 2735
Formula Weight 241.46
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Hexyldecan-1-amine