accessibility menu, dialog, popup

UserName

CAS RN 62281-05-4

IUPAC Name: 7-(aminomethyl)pentadecane
Synonyms: 7-(aminomethyl)pentadecane
Molecular Weight (g/mol): 241.46
Molecular Formula: C16H35N
InChi Key: HEGUXAZWCQVLPV-UHFFFAOYNA-N
SMILES: CCCCCCCCC(CN)CCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results