missing translation for 'onlineSavingsMsg'
Learn More

2-Hexyl-6-phenylpyridine 98.0+%, TCI America™

Supplier:  TCI America H12651G

Catalog No. H12651G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Hexyl-6-phenylpyridine
Yellow
1 g
CCCCCCC1=CC=CC(=N1)C2=CC=CC=C2
239.362
239.36
Liquid
499158-97-3
C17H21N
CIPJZUAHRIZGJN-UHFFFAOYSA-N
2-hexyl-6-phenylpyridine
44629964
≥98.0% (GC,T)

Chemical Identifiers

499158-97-3
239.362
44629964
CCCCCCC1=CC=CC(=N1)C2=CC=CC=C2
C17H21N
CIPJZUAHRIZGJN-UHFFFAOYSA-N
2-hexyl-6-phenylpyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions