CAS RN 499158-97-3
CAS RN 499158-97-3
IUPAC Name:
2-hexyl-6-phenylpyridine
Synonyms:
2-hexyl-6-phenylpyridine
Molecular Weight (g/mol):
239.36
Molecular Formula:
C17H21N
InChi Key:
CIPJZUAHRIZGJN-UHFFFAOYSA-N
SMILES:
CCCCCCC1=CC=CC(=N1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Hexyl-6-phenylpyridine 98.0+%, TCI America™
CAS: 499158-97-3 Molecular Formula: C17H21N Molecular Weight (g/mol): 239.362 InChI Key: CIPJZUAHRIZGJN-UHFFFAOYSA-N PubChem CID: 44629964 IUPAC Name: 2-hexyl-6-phenylpyridine SMILES: CCCCCCC1=CC=CC(=N1)C2=CC=CC=C2