missing translation for 'onlineSavingsMsg'
Learn More

2-Hexanone, 98%

CAS: 591-78-6 | C6H12O | 100.16 g/mol

Supplier:  Thermo Scientific Chemicals A1486718

 View more versions of this product

Catalog No. AAA1486718


Only null left
Explore available promotions

Description

Description

2-Hexanone acts as a solvent for lacquers, ink thinners, nitrocellulose, resins and waxes. It is also employed in the preparation of (±)-monomorine. It also plays a useful role in the preparation of alcohols by transfer hydrogenation of ketones. Further, it is used as a medium evaporating solvent for nitrocellulose acrylates, vinyl and alkyd coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hexanone acts as a solvent for lacquers, ink thinners, nitrocellulose, resins and waxes. It is also employed in the preparation of (+/-)-monomorine. It also plays a useful role in the preparation of alcohols by transfer hydrogenation of ketones. Further, it is used as a medium evaporating solvent for nitrocellulose acrylates, vinyl and alkyd coatings.

Solubility
Miscible with acetone, ethanol and ether. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases and reducing agents.
Specifications

Specifications

2-Hexanone
-56°C
126°C to 127°C
Pungent
1.401
50 g
1737676
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
QQZOPKMRPOGIEB-UHFFFAOYSA-N
hexan-2-one
11583
98%
591-78-6
0.811
23°C (73°F)
C6H12O
MFCD00009482
UN1224
14,6033
Miscible with acetone,ethanol and ether. Slightly miscible with water.
CCCCC(C)=O
100.16
100.16

Chemical Identifiers

591-78-6
100.16
QQZOPKMRPOGIEB-UHFFFAOYSA-N
11583
CCCCC(C)=O
C6H12O
MFCD00009482
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
hexan-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H372-H361-H226-H336
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapour.
May cause drowsiness or dizziness.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P308+P313-P370+P378q-P501c

H226-H336-H361f-H372

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 209-731-1

RTECSNumber : MP1400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only