missing translation for 'onlineSavingsMsg'
Learn More

2-Hexanone, 98%

Catalog No. aaa1486736
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1486736 500 g
AAA1486709 10 g
AAA1486718 50 g
AAA1486722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1486736 Supplier Thermo Scientific Chemicals Supplier No. A1486736
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 591-78-6 | C6H12O | 100.16 g/mol

2-Hexanone acts as a solvent for lacquers, ink thinners, nitrocellulose, resins and waxes. It is also employed in the preparation of (±)-monomorine. It also plays a useful role in the preparation of alcohols by transfer hydrogenation of ketones. Further, it is used as a medium evaporating solvent for nitrocellulose acrylates, vinyl and alkyd coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hexanone acts as a solvent for lacquers, ink thinners, nitrocellulose, resins and waxes. It is also employed in the preparation of (+/-)-monomorine. It also plays a useful role in the preparation of alcohols by transfer hydrogenation of ketones. Further, it is used as a medium evaporating solvent for nitrocellulose acrylates, vinyl and alkyd coatings.

Solubility
Miscible with acetone, ethanol and ether. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases and reducing agents.

Chemical Identifiers

CAS 591-78-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009482
InChI Key QQZOPKMRPOGIEB-UHFFFAOYSA-N
Synonym 2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
PubChem CID 11583
IUPAC Name hexan-2-one
SMILES CCCCC(C)=O

Specifications

Melting Point -56°C
Density 0.811
Boiling Point 126°C to 127°C
Flash Point 23°C (73°F)
Odor Pungent
Refractive Index 1.401
Quantity 500 g
UN Number UN1224
Beilstein 1737676
Merck Index 14,6033
Solubility Information Miscible with acetone,ethanol and ether. Slightly miscible with water.
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material 2-Hexanone
Show More Show Less
Hazard Category H226-H336-H361f-H372
Hazard Statement GHS H Statement
H372-H361-H226-H336
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapour.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 209-731-1
RTECSNumber MP1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only