missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Hexanol, 99%

Catalog No. p-7044351
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0599314 25 g
AAL0599322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0599314 Supplier Thermo Scientific Chemicals Supplier No. L0599314
Only null left

CAS: 626-93-7 | C6H14O | 102.177 g/mol

(±)-2-Hexanol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff field. It is used as perfuming agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-2-Hexanol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff field. It is used as perfuming agent.

Solubility
Soluble in water (13 g/L at 25°C),

Notes
Incompatible with acid chlorides and oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 626-93-7
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004585
InChI Key QNVRIHYSUZMSGM-UHFFFAOYSA-N
Synonym 2-hexanol, sec-hexyl alcohol, 2-hydroxyhexane, 2-hexyl alcohol, n-butylmethylcarbinol, 2-hexanol, r, 2-hexanol, s, n-c4h9ch oh ch3, methylamyl alcohol, sec-hexanol
PubChem CID 12297
IUPAC Name hexan-2-ol
SMILES CCCCC(C)O

Specifications

Melting Point -23°C
Density 0.81
Boiling Point 137°C to 139°C
Flash Point 45°C (113°F)
Odor Characteristic
Refractive Index 1.414
Quantity 25 g
UN Number UN2282
Beilstein 1718996
Solubility Information Soluble in water (13g/L at 25°C),
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material (+/-)-2-Hexanol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXANOLS
EINECSNumber 210-971-4
RTECSNumber MO8470000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only