missing translation for 'onlineSavingsMsg'
Learn More

2-Hexanol, 99%

Catalog No. AC156290500
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156290500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC156290500 Supplier Thermo Scientific Chemicals Supplier No. 156290500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 626-93-7 | C6H14O | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 626-93-7
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004585
InChI Key QNVRIHYSUZMSGM-UHFFFAOYSA-N
Synonym 2-hexanol, sec-hexyl alcohol, 2-hydroxyhexane, 2-hexyl alcohol, n-butylmethylcarbinol, 2-hexanol, r, 2-hexanol, s, n-c4h9ch oh ch3, methylamyl alcohol, sec-hexanol
PubChem CID 12297
IUPAC Name hexan-2-ol
SMILES CCCCC(C)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -23.0°C
Color Colorless
Density 0.8100g/mL
Boiling Point 139.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4125 to 1.4155
Linear Formula CH3(CH2)3CH(OH)CH3
Quantity 50 g
Beilstein 01, 408
Specific Gravity 0.81
Solubility Information Solubility in water: insoluble.
Formula Weight 102.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Hexanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 210-971-4

RUO – Research Use Only