missing translation for 'onlineSavingsMsg'
Learn More

2-Heptyn-1-ol, 97%

Catalog No. AAA1495903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1495903 1 g
AAA1495906 5 g
AAA1495914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1495903 Supplier Thermo Scientific Chemicals Supplier No. A1495903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1002-36-4 | C7H12O | 112.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1002-36-4
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00039541
InChI Key OGDYROFIIXDTQK-UHFFFAOYSA-N
Synonym 2-heptyn-1-ol, 2-heptyne-1-ol, 2-heptynol, pubchem13065, acmc-2097no, hoch2c=c ch2 3ch3
PubChem CID 535111
IUPAC Name hept-2-yn-1-ol
SMILES CCCCC#CCO

Specifications

Density 0.882
Boiling Point 89°C to 90°C (16 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.454
Quantity 1 g
Beilstein 1739001
Formula Weight 112.17
Percent Purity 97%
Chemical Name or Material 2-Heptyn-1-ol
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only