accessibility menu, dialog, popup

UserName

CAS RN 1002-36-4

IUPAC Name: hept-2-yn-1-ol
Synonyms: hept-2-yn-1-ol
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: OGDYROFIIXDTQK-UHFFFAOYSA-N
SMILES: CCCCC#CCO
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)