missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Heptylcyclopentanone 99.0+%, TCI America™
$504.39 - $504.39
Chemical Identifiers
| CAS | 137-03-1 |
|---|---|
| Molecular Formula | C12H22O |
| Molecular Weight (g/mol) | 182.31 |
| MDL Number | MFCD00019315 |
| InChI Key | PJXHBTZLHITWFX-UHFFFAOYNA-N |
| Synonym | 2-heptylcyclopentanone, 2-n-heptylcyclopentanone, cyclopentanone, 2-heptyl, alismone, cyclopentanone, 2-n-heptyl, alpha-heptyl cyclopentanone, alpha-heptylcyclopentanone, 2-n-heptyl cyclopentanone, heptylcyclopentan-1-one, heptylcyclopentanone |
| PubChem CID | 8710 |
| IUPAC Name | 2-heptylcyclopentan-1-one |
| SMILES | CCCCCCCC1CCCC1=O |
Chemical Identifiers
| 137-03-1 | |
| 182.31 | |
| PJXHBTZLHITWFX-UHFFFAOYNA-N | |
| 8710 | |
| CCCCCCCC1CCCC1=O |
| C12H22O | |
| MFCD00019315 | |
| 2-heptylcyclopentanone, 2-n-heptylcyclopentanone, cyclopentanone, 2-heptyl, alismone, cyclopentanone, 2-n-heptyl, alpha-heptyl cyclopentanone, alpha-heptylcyclopentanone, 2-n-heptyl cyclopentanone, heptylcyclopentan-1-one, heptylcyclopentanone | |
| 2-heptylcyclopentan-1-one |
Specifications
| 137-03-1 | |
| 264°C | |
| MFCD00019315 | |
| 2-heptylcyclopentanone, 2-n-heptylcyclopentanone, cyclopentanone, 2-heptyl, alismone, cyclopentanone, 2-n-heptyl, alpha-heptyl cyclopentanone, alpha-heptylcyclopentanone, 2-n-heptyl cyclopentanone, heptylcyclopentan-1-one, heptylcyclopentanone | |
| CCCCCCCC1CCCC1=O | |
| 182.31 | |
| 182.31 | |
| Liquid |
| Colorless | |
| C12H22O | |
| 25 mL | |
| PJXHBTZLHITWFX-UHFFFAOYNA-N | |
| 2-heptylcyclopentan-1-one | |
| 8710 | |
| ≥99.0% (GC) | |
| 2-Heptylcyclopentanone |
Safety and Handling
RTECSNumber : GY4950000
TSCA : Yes