accessibility menu, dialog, popup

UserName

CAS RN 137-03-1

H3CO

CAS RN 137-03-1

IUPAC Name: 2-heptylcyclopentan-1-one
Synonyms: 2-heptylcyclopentanone
Molecular Weight (g/mol): 182.31
Molecular Formula: C12H22O
InChi Key: PJXHBTZLHITWFX-UHFFFAOYNA-N
SMILES: CCCCCCCC1CCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results