Learn More
2-Heptanone, 98%
CAS: 110-43-0 | C7H14O | 114.19 g/mol
Supplier: Thermo Scientific Chemicals 154000010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Heptanone | |
| 97.5 | |
| -35.0°C | |
| 0.8200g/mL | |
| 39°C | |
| 98% | |
| C7H14O | |
| CH3(CH2)4COCH3 | |
| 1 L | |
| 0.82 | |
| 2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone | |
| CATSNJVOTSVZJV-UHFFFAOYSA-N | |
| heptan-2-one | |
| 8051 | |
| 114.19 | |
| Pure |
| 110-43-0 | |
| 100.0 | |
| Colorless | |
| 149.0°C to 150.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4075 to 1.4095 | |
| MFCD00009513 | |
| 01, 699 | |
| 15, 4698 | |
| Solubility in water: 4.3g/L (20°C). Other solubilities: soluble in alcohol and ether | |
| CCCCCC(C)=O | |
| 114.19 | |
| CHEBI:5672 | |
| 98% | |
| Liquid |
Chemical Identifiers
| 110-43-0 | |
| 114.19 | |
| CATSNJVOTSVZJV-UHFFFAOYSA-N | |
| 8051 | |
| heptan-2-one |
| C7H14O | |
| MFCD00009513 | |
| 2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone | |
| CHEBI:5672 | |
| CCCCCC(C)=O |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
May cause drowsiness or dizziness.
Flammable liquid and vapor.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Warning
EINECSNumber : 203-767-1
RUO – Research Use Only