missing translation for 'onlineSavingsMsg'
Learn More

2-Furoic Acid, 97-100%, Spectrum™ Chemical

Catalog No. p-7165569
Change view
Click to view available options
Quantity:
100 g
25 kg
Packaging:
Fiber Drum
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608217 100 g Poly Bottle
18608218 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18608217 Supplier Spectrum Chemical Mfg Cor Supplier No. F1140100GM
Only null left

Chemical Identifiers

CAS 88-14-2

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 100 g
Percent Purity 97 to 100%
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.