missing translation for 'onlineSavingsMsg'
Learn More

2-Furancarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7129488
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F008125G 25 g
F0081500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F008125G Supplier TCI America Supplier No. F008125G
Only null left

Chemical Identifiers

CAS 88-14-2
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.084
MDL Number MFCD00003238
InChI Key SMNDYUVBFMFKNZ-UHFFFAOYSA-N
Synonym 2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoova
PubChem CID 6919
ChEBI CHEBI:30845
IUPAC Name furan-2-carboxylic acid
SMILES C1=COC(=C1)C(=O)O

Specifications

Melting Point 132°C
Color White-Yellow
Boiling Point 232°C
Quantity 25 g
Formula Weight 112.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Furancarboxylic Acid
EINECSNumber (5)-6921
RTECSNumber LV1763000
TSCA Yes