missing translation for 'onlineSavingsMsg'
Learn More

2-Furaldehyde 2,2-dimethylhydrazone, 98%

Catalog No. AAH3201309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3201309 10 g
AAH3201318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3201309 Supplier Thermo Scientific Chemicals Supplier No. H3201309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14064-21-2 | C7H10N2O | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14064-21-2
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD00010685,MFCD00010685
InChI Key DURWBNUASAZMSN-SOFGYWHQSA-N
Synonym e-2-furan-2-ylmethylidene-1,1-dimethylhydrazine, furfural dimethyl hydrazone
PubChem CID 5372420
SMILES CN(C)\N=C\C1=CC=CO1

Specifications

Density 1.048
Boiling Point 69°C (2 mmHg)
Flash Point 92°C (198°F)
Odor Recognizable
Refractive Index 1.58
Quantity 10 g
IUPAC Name N-[(Z)-furan-2-ylmethylideneamino]-N-methylmethanamine
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 2-Furaldehyde 2,2-dimethylhydrazone
Hazard Category H227-H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only