accessibility menu, dialog, popup

UserName

CAS RN 14064-21-2

IUPAC Name: (2E)-2-[(furan-2-yl)methylidene]-1,1-dimethylhydrazine
Synonyms: (2E)-2-(furan-2-ylmethylidene)-1,1-dimethylhydrazine
Molecular Weight (g/mol): 138.17
Molecular Formula: C7H10N2O
InChi Key: DURWBNUASAZMSN-SOFGYWHQSA-N
SMILES: CN(C)\N=C\C1=CC=CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results