missing translation for 'onlineSavingsMsg'
Learn More

2-Formylthiazole 97.0+%, TCI America™

Catalog No. p-7129601
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
F03031G 1 g
F03035G 5 g
2 options

Catalog No. F03031G

Supplier: TCI America F03031G

Only null left

Chemical Identifiers

CAS 10200-59-6
Molecular Formula C4H3NOS
Molecular Weight (g/mol) 113.134
MDL Number MFCD00142924
InChI Key ZGTFNNUASMWGTM-UHFFFAOYSA-N
Synonym 2-thiazolecarboxaldehyde, thiazole-2-carbaldehyde, 2-formylthiazole, thiazole-2-carboxaldehyde, 2-formyl thiazole, thiazolecarboxaldehyde, 2-formyl-1,3-thiazole, 1,3-thiazole-2-carboxaldehyde, 2-thiazolecarbaldehyde, pubchem9929
PubChem CID 2734903
ChEBI CHEBI:43623
IUPAC Name 1,3-thiazole-2-carbaldehyde
SMILES C1=CSC(=N1)C=O

Specifications

Color Yellow
Boiling Point 70°C
Quantity 1 g
Formula Weight 113.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Formylthiazole
TSCA No
Recommended Storage Refrigerator