missing translation for 'onlineSavingsMsg'
Learn More

2-Formylbenzonitrile 99.0+%, TCI America™

Catalog No. p-7124425
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C04445G 5 g
C044425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C04445G Supplier TCI America Supplier No. C04445G

Chemical Identifiers

CAS 7468-67-9
Molecular Formula C8H5NO
Molecular Weight (g/mol) 131.13
MDL Number MFCD00017503
InChI Key QVTPWONEVZJCCS-UHFFFAOYSA-N
Synonym 2-cyanobenzaldehyde, o-cyanobenzaldehyde, benzonitrile, 2-formyl, cyanobenzaldehyde, 2-formylbenzenecarbonitrile, 2-cyano-benzaldehyde, 2-formyl-benzonitrile, pubchem7409, #, acmc-1blgc
PubChem CID 101209
IUPAC Name 2-formylbenzonitrile
SMILES O=CC1=CC=CC=C1C#N

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 131.13
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Formylbenzonitrile