missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Formylbenzenesulfonic acid, sodium salt, 90%, tech.
CAS: 1008-72-6 | C7H5NaO4S | 208.16 g/mol
$319.97 - $319.97
Chemical Identifiers
| CAS | 1008-72-6 |
|---|---|
| Molecular Formula | C7H5NaO4S |
| Molecular Weight (g/mol) | 208.16 |
| MDL Number | MFCD00007478 |
| InChI Key | ADPUQRRLAAPXGT-UHFFFAOYSA-M |
| Synonym | sodium 2-formylbenzenesulfonate, 2-formylbenzenesulfonic acid sodium salt, sodium o-formylbenzenesulfonate, benzenesulfonic acid, 2-formyl-, sodium salt, unii-lh8xh59068, 2-sulfobenzaldehyde sodium salt, sodium benzaldehyde-2-sulfonate, benzaldehyde-2-sulfonic acid sodium salt, o-formylbenzenesulfonic acid sodium salt, sodium 2-formylbenzene-1-sulfonate |
| PubChem CID | 3794540 |
| IUPAC Name | sodium;2-formylbenzenesulfonate |
| SMILES | [Na+].[O-]S(=O)(=O)C1=CC=CC=C1C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC219325000
|
Thermo Scientific Chemicals
219325000 |
500 g | Plastic bottle |
Each for $319.97
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1008-72-6 | |
| 208.16 | |
| ADPUQRRLAAPXGT-UHFFFAOYSA-M | |
| 3794540 | |
| [Na+].[O-]S(=O)(=O)C1=CC=CC=C1C=O |
| C7H5NaO4S | |
| MFCD00007478 | |
| sodium 2-formylbenzenesulfonate, 2-formylbenzenesulfonic acid sodium salt, sodium o-formylbenzenesulfonate, benzenesulfonic acid, 2-formyl-, sodium salt, unii-lh8xh59068, 2-sulfobenzaldehyde sodium salt, sodium benzaldehyde-2-sulfonate, benzaldehyde-2-sulfonic acid sodium salt, o-formylbenzenesulfonic acid sodium salt, sodium 2-formylbenzene-1-sulfonate | |
| sodium;2-formylbenzenesulfonate |
Specifications
| 1008-72-6 | |
| 100.0 | |
| Authentic | |
| Plastic bottle | |
| HCOC6H4SO3Na | |
| 500 g | |
| sodium 2-formylbenzenesulfonate, 2-formylbenzenesulfonic acid sodium salt, sodium o-formylbenzenesulfonate, benzenesulfonic acid, 2-formyl-, sodium salt, unii-lh8xh59068, 2-sulfobenzaldehyde sodium salt, sodium benzaldehyde-2-sulfonate, benzaldehyde-2-sulfonic acid sodium salt, o-formylbenzenesulfonic acid sodium salt, sodium 2-formylbenzene-1-sulfonate | |
| [Na+].[O-]S(=O)(=O)C1=CC=CC=C1C=O | |
| 208.16 | |
| 208.17 | |
| Technical | |
| 2-Formylbenzenesulfonic acid sodium salt, tech., 90% |
| 88.0 | |
| White to Yellow | |
| 90% | |
| C7H5NaO4S | |
| MFCD00007478 | |
| 11, 324 | |
| ADPUQRRLAAPXGT-UHFFFAOYSA-M | |
| sodium;2-formylbenzenesulfonate | |
| 3794540 | |
| 90% | |
| Powder |
Safety and Handling
AIR SENSITIVE
EINECSNumber : 213-758-4
TSCA : TSCA
RUO – Research Use Only