missing translation for 'onlineSavingsMsg'
Learn More

2-Formylbenzenesulfonic acid, sodium salt, 90%, tech.

Catalog No. AC219325000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219325000 500 g Plastic bottle
AC219320025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC219325000 Supplier Thermo Scientific Chemicals Supplier No. 219325000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1008-72-6 | C7H5NaO4S | 208.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1008-72-6
Molecular Formula C7H5NaO4S
Molecular Weight (g/mol) 208.16
MDL Number MFCD00007478
InChI Key ADPUQRRLAAPXGT-UHFFFAOYSA-M
Synonym sodium 2-formylbenzenesulfonate, 2-formylbenzenesulfonic acid sodium salt, sodium o-formylbenzenesulfonate, benzenesulfonic acid, 2-formyl-, sodium salt, unii-lh8xh59068, 2-sulfobenzaldehyde sodium salt, sodium benzaldehyde-2-sulfonate, benzaldehyde-2-sulfonic acid sodium salt, o-formylbenzenesulfonic acid sodium salt, sodium 2-formylbenzene-1-sulfonate
PubChem CID 3794540
IUPAC Name sodium;2-formylbenzenesulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=CC=C1C=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Plastic bottle
Linear Formula HCOC6H4SO3Na
Quantity 500 g
Beilstein 11, 324
Formula Weight 208.17
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material 2-Formylbenzenesulfonic acid sodium salt, tech., 90%
Show More Show Less
Hazard Category AIR SENSITIVE
EINECSNumber 213-758-4
TSCA TSCA

RUO – Research Use Only